accessibility menu, dialog, popup

UserName

CH3NHH3CHN

CAS RN 108-49-6

IUPAC Name: 2,6-dimethylpiperazine
Synonyms: 2,6-dimethylpiperazine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: IFNWESYYDINUHV-UHFFFAOYNA-N
SMILES: CC1CNCC(C)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results