CAS RN 107767-58-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
A-802715, MedChemExpress
MedChemExpress A802715 is a methylxanthine derivative. A802715 has a TD50 (toxic dose of 50%) of 0.9-1.1 mM.