CAS RN 107485-64-3
CAS RN 107485-64-3
IUPAC Name:
2-chloro-6-nitrobenzamide
Synonyms:
2-chloro-6-nitrobenzamide
Molecular Weight (g/mol):
200.58
Molecular Formula:
C7H5ClN2O3
InChi Key:
WPVXBESIHVJEOI-UHFFFAOYSA-N
SMILES:
NC(=O)C1=C(Cl)C=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
2-Chloro-6-nitrobenzamide, 98%, Thermo Scientific™
CAS: 107485-64-3 Molecular Formula: C7H5ClN2O3 Molecular Weight (g/mol): 200.578 MDL Number: MFCD02093931 InChI Key: WPVXBESIHVJEOI-UHFFFAOYSA-N PubChem CID: 86279 IUPAC Name: 2-chloro-6-nitrobenzamide SMILES: C1=CC(=C(C(=C1)Cl)C(=O)N)[N+](=O)[O-]