CAS RN 107481-28-7
IUPAC Name:
9-(prop-2-enoyloxy)nonyl prop-2-enoate
Synonyms:
9-(prop-2-enoyloxy)nonyl prop-2-enoate
Molecular Weight (g/mol):
268.35
Molecular Formula:
C15H24O4
InChi Key:
PGDIJTMOHORACQ-UHFFFAOYSA-N
SMILES:
C=CC(=O)OCCCCCCCCCOC(=O)C=C
1
–
3
of
3
results
1
1,9-Nonanediol Diacrylate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,9-Nonanediol Diacrylate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,9-Nonanediol Diacrylate (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.