man

Test

CAS RN 107445-21-6

H3CNHNO

CAS RN 107445-21-6

IUPAC Name: 4-(1-nitrosoethylidene)-1,4-dihydropyridine
Synonyms: 4-(1-nitrosoethylidene)-1H-pyridine
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: GROZUESHXQOJFW-UHFFFAOYSA-N
SMILES: CC(N=O)=C1C=CNC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results