CAS RN 1070660-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Ethylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester, TRC
CAS: 1070660-34-2 Molecular Formula: C13 H20 N2 O2 Molecular Weight (g/mol): 236.31 Synonym: Carbamic acid, N-ethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester,3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ethyl Carbamate,N-Ethylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester IUPAC Name: [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethylcarbamate SMILES: CCNC(=O)Oc1cccc(c1)[C@H](C)N(C)C