CAS RN 107-64-2
IUPAC Name:
dimethyldioctadecylazanium chloride
Synonyms:
distearyldimonium chloride dimethyl dioctadecylammonium chloride
Molecular Weight (g/mol):
586.52
Molecular Formula:
C38H80ClN
InChi Key:
REZZEXDLIUJMMS-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
1
–
3
of
3
results
1
Dimethyldioctadecylammonium Chloride (mixture of C16 to C18 chain lengths), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dimethyldioctadecylammonium Chloride (mixture of C16 to C18 chain lengths), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dimethyldioctadecylammonium Chloride (mixture of C16 to C18 chain lengths), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.