accessibility menu, dialog, popup

UserName

H3COPSOH3CSSOCH3CH3H3C

CAS RN 10548-10-4

IUPAC Name: O,O-diethyl {[(2-methylpropane-2-sulfinyl)methyl]sulfanyl}phosphonothioate
Synonyms: terbufos sulfoxide
Molecular Weight (g/mol): 304.42
Molecular Formula: C9H21O3PS3
InChi Key: KKOYBOFPUBHPFO-UHFFFAOYNA-N
SMILES: CCOP(=S)(OCC)SCS(=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results