CAS RN 1049718-43-5
CAS RN 1049718-43-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Deschloro-3-bromo Bupropion Hydcrochloride, TRC
CAS: 1049718-43-5 Molecular Formula: C13 H18 Br N O . Cl H Molecular Weight (g/mol): 320.65 Synonym: 2-(tert-Butylamino)-3'-bromopropiophenone Hydrochloride IUPAC Name: 1-(3-bromophenyl)-2-(tert-butylamino)propan-1-one;hydrochloride SMILES: Cl.CC(NC(C)(C)C)C(=O)c1cccc(Br)c1