CAS RN 10429-30-8
IUPAC Name:
2-[3-(dimethylamino)propoxy]benzaldehyde
Synonyms:
2-[3-(dimethylamino)propoxy]benzaldehyde
Molecular Weight (g/mol):
207.27
Molecular Formula:
C12H17NO2
InChi Key:
DSHCOEWHRLVOTF-UHFFFAOYSA-N
SMILES:
CN(C)CCCOC1=CC=CC=C1C=O
1
–
1
of
1
results
1
2-[3-(Dimethylamino)propoxy]benzaldehyde, 97%, Thermo Scientific™
CAS: 10429-30-8 Molecular Formula: C12H17NO2 Molecular Weight (g/mol): 207.27 MDL Number: MFCD00128460 InChI Key: DSHCOEWHRLVOTF-UHFFFAOYSA-N Synonym: 2-3-dimethylamino propoxy benzaldehyde,2-3-dimethylamino-propoxy-benzaldehyde,2-3-dimethylaminopropoxy benzaldehyde,2-3-dimethylaminopropoxy-benzaldehyde,acmc-1c25w,2-3-dimethylaminopropoxy bezaldehyde,2-3-dimethylaminopropoxy benzal-dehyde,2-3-dimethylaminopropoxyl benzaldehyde,2-3-dimethylamino-propoxy benzaldehyde PubChem CID: 2995968 IUPAC Name: 2-[3-(dimethylamino)propoxy]benzaldehyde SMILES: CN(C)CCCOC1=CC=CC=C1C=O