accessibility menu, dialog, popup

UserName

CAS RN 1029439-56-2

BOOH3CCH3H3CH3CNH

CAS RN 1029439-56-2

IUPAC Name: N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}aniline
Synonyms: N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}aniline
Molecular Weight (g/mol): 309.22
Molecular Formula: C19H24BNO2
InChi Key: KNYGNGFJIPTYBJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CNC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results