accessibility menu, dialog, popup

UserName

CAS RN 10203-32-4

H3COHCH3

CAS RN 10203-32-4

IUPAC Name: dodecan-4-ol
Synonyms: dodecan-4-ol
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: ZGSIAHIBHSEKPB-UHFFFAOYNA-N
SMILES: CCCCCCCCC(O)CCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)