accessibility menu, dialog, popup

UserName

CAS RN 1020253-79-5

H3COPNCH3CH3CH3H3CNCH3H3CCH3CH3

CAS RN 1020253-79-5

IUPAC Name: {[bis(propan-2-yl)amino](propoxy)phosphanyl}bis(propan-2-yl)amine
Synonyms: [(diisopropylamino)(propoxy)phosphanyl]diisopropylamine
Molecular Weight (g/mol): 290.43
Molecular Formula: C15H35N2OP
InChi Key: OTEPWYBGBKUVGE-UHFFFAOYSA-N
SMILES: CCCOP(N(C(C)C)C(C)C)N(C(C)C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results