accessibility menu, dialog, popup

UserName

CAS RN 1017779-57-5

IUPAC Name: 4-ethoxy-2,3-difluorobenzamide
Synonyms: 4-ethoxy-2,3-difluorobenzamide
Molecular Weight (g/mol): 201.17
Molecular Formula: C9H9F2NO2
InChi Key: PFXUEERRMBKGOD-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=C(C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results