accessibility menu, dialog, popup

UserName

CAS RN 1017779-30-4

CH3FFFNH2

CAS RN 1017779-30-4

IUPAC Name: 1-[5-methyl-2-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[5-methyl-2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 189.18
Molecular Formula: C9H10F3N
InChi Key: LDYZJNWJFZYWKD-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results