accessibility menu, dialog, popup

UserName

CAS RN 1017778-84-5

H3COFFFOOH

CAS RN 1017778-84-5

IUPAC Name: 2-[2-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: LMQFQFJTFHHNQH-UHFFFAOYSA-N
SMILES: COC1=C(CC(O)=O)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results