accessibility menu, dialog, popup

UserName

FClClOHO

CAS RN 10129-77-8

IUPAC Name: 2-(2,4-dichloro-5-fluorophenyl)acetic acid
Synonyms: (2,4-dichloro-5-fluorophenyl)acetic acid
Molecular Weight (g/mol): 223.02
Molecular Formula: C8H5Cl2FO2
InChi Key: LFLORXCBEGTRHI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(F)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results