accessibility menu, dialog, popup

UserName

PbOVOOOVOO

CAS RN 10099-79-3

IUPAC Name: λ²-lead(2+) divanadiumoylolate
Synonyms: λ²-lead(2+) bis(dioxidovanadium(1+))
Molecular Weight (g/mol): 405.1
Molecular Formula: O6PbV2
InChi Key: OGRLITDAVSILTM-UHFFFAOYSA-N
SMILES: [Pb++].[O-][V](=O)=O.[O-][V](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results