accessibility menu, dialog, popup

UserName

FFCr

CAS RN 10049-10-2

IUPAC Name: λ²-chromium(2+) difluoride
Synonyms: chromium(II) fluoride
Molecular Weight (g/mol): 89.99
Molecular Formula: CrF2
InChi Key: RNFYGEKNFJULJY-UHFFFAOYSA-L
SMILES: [F-].[F-].[Cr++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results