accessibility menu, dialog, popup

UserName

CAS RN 100348-93-4

CAS RN 100348-93-4

IUPAC Name: 8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione
Synonyms: wieland-miescher ketone
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: DNHDRUMZDHWHKG-UHFFFAOYNA-N
SMILES: CC12CCC(=O)C=C1CCCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results