accessibility menu, dialog, popup

UserName

CAS RN 1000017-97-9

H3COONNNH2

CAS RN 1000017-97-9

IUPAC Name: ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 205.22
Molecular Formula: C10H11N3O2
InChi Key: SDHJUHBOHSJBEH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C(C=CC=C2N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results