missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZOOM™ Urea

Product Code. 10042532
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10042532 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10042532 Supplier Invitrogen™ Supplier No. ZU10001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The ZOOM™ IEF Fractionator offers a fast, reliable method to reduce sample complexity, enrich low abundance proteins, and increase the dynamic range of detection.

The ZOOM™ IEF Fractionator offers a fast, reliable method to reduce sample complexity, enrich low abundance proteins, and increase the dynamic range of detection. Solution phase isoelectric focusing with the ZOOM™ IEF Fractionator provides reproducible separations in three hours. Fractionated samples are ready for further analysis by two dimensional gel electrophoresis (2DE), one dimensional gel electrophoresis (1DE), or two dimensional liquid chromatography/mass spectrometry (2D LC/MS).

The ZOOM™ IEF Fractionator System (Figure 1) consists of the following components:

  • ZOOM™ IEF Fractionator is built for safety, reliability, and durability. It includes easy-to-assemble sample chambers, in-built buffer chambers, buffer trough, removable electrode assembly, and safety lid.
  • ZOOM™ Disks are pre-cast polyacrylamide gels that eliminate the need for manual preparation and minimize any chance of cross contamination. These immobilized buffered disks are pre-labeled, disposable, and designed for single use, ensuring consistent and reproducible fractionation. They resolve samples into as many as 7 fractions (using 8 disks of specific pH) to six fractions (using six disks of specific pH) from pH 3-12. Simply place each disk between the sample chambers in the ZOOM™ IEF Fractionator to allow separation in a specific pH range.
  • ZOOM™ Reagents are proteomic grade reagents, including ZOOM™ 2D Protein Solubilizers urea, thiourea, CHAPS, and ampholytes, that ensure optimal conditions for sample preparation.
  • Basic Protein Kit which includes ZOOM™ Discs and ZOOM Strips for IEF of basic proteins, focusing buffers, and solubilizer kits.

Sample Chamber and Fractionator Unit Set-up Procedure

Figures 4-6 illustrate assembly of the sample chambers and fractionator unit. The sample chamber and components fit together easily and offer a leak-proof seal. Simply place disks between the sample chambers in the ZOOM™ IEF Fractionator to allow separation in a specific pH range. For example, if you want to fractionate proteins between pH 4.6 and 5.4, flank a sample chamber with a ZOOM (r) Disk of pH 4.6 and one of pH 5.4.

Downstream Processing by 2D Gel Electrophoresis

Following fractionation, the separated samples can be subjected to further analysis using narrow or broad range ZOOM™ Strips on the ZOOM™ IPGRunner™ System. Follow with second dimension analysis using neutral pH NuPAGE™ ZOOM™ Gels.

TRUSTED_SUSTAINABILITY

Specifications

Description ZOOM™ Urea, 1kg, White, Powder, 219°C Melting Point, Water Soluble, Room Temperature Shipping Condition, For Research use Only, Used in Protein Gel Electrophoresis, Protein Purification and Isolation
Buffer Anode and cathode buffer
Color White
Content And Storage ZOOM™ IEF Fractionator
• ZOOM™ Disks, one pack of each pH

ZOOM™ Reagents
• Urea
• Thiourea
• CHAPS
• Carrier ampholytes pH 3-10
• Anode buffers
• Bathode buffers

Store all components at 4°C.
Shipping Condition Room Temperature
Product Line ZOOM
Form Powder
Formulation Powder
For Use With (Application) Protein gel electrophoresis, protein purification and isolation, isoelectric focusing, 2D gel electrophoresis, protein biology
Reagent Type Urea
Quantity 1 kg
Kit Contents ZOOM™ IEF fractionator; one pack ZOOM™ disks of each pH; and ZOOM™ Reagents ZOOM™ disks, carrier ampholytes and buffers
Product Type Urea
Melting Temperature 219°C
Storage Requirements 4°C
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.