Learn More
Emma Safety Footwear Zion Safety Shoes
Product Code. 15998291
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15998291 46 1 pair
15908291 37 1 pair
15918291 38 1 pair
15928291 39 1 pair
15938291 40 1 pair
15948291 41 1 pair
15958291 42 1 pair
15968291 43 1 pair
15978291 44 1 pair
15988291 45 1 pair
15908301 47 1 pair
15918301 48 1 pair
15928301 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15998291 Supplier Emma Safety Footwear Supplier No. MM30084846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical resistant with highly visible 3M™ reflective strip. The impenetrable sole protects feet from sharp and metal objects while the steel toe cap and reinforced polyurethane toe cover offer protection from falling objects. Microfiber components stand up to cement and remains flexible.
  • S3-Safety shoe with sole plate for wet work
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Black
Design Zion
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 46