missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99%, (trace metal basis), < 10 microns

Product Code. 10076143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076143 25 g Glass bottle 25g
10667212 100 g Glass bottle 100g
10425485 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10076143 Supplier Thermo Scientific Acros Supplier No. 223650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-98-3 | SZn | 97.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99%
Linear Formula ZnS
Merck Index 15, 10360
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
IUPAC Name sulfanylidenezinc
Formula Weight 97.44
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Zinc sulfide, 99.99%, <10 microns
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.