missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™

Product Code. p-7120917 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
7 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
100g
500g
7kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15612280 100 g Poly Bottle 100g
15622280 500 g Poly Bottle 500g
15632280 7 kg Drum with inliner 7kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15612280 Supplier Honeywell Research Chemicals Supplier No. 228737100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 38.9°C
Color White
Quantity 100 g
Linear Formula Zn(NO3)2 · 6H2O
UN Number UN1514
Solubility Information Soluble in water
IUPAC Name zinc;dinitrate;hexahydrate
Formula Weight 297.49g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Powder or Crystals or Flakes
Density 2.060g/cm3
Chemical Name or Material Zinc Nitrate Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.