Learn More
Usbeck Laborgeraete™ Zinc Die-casting Stand Clamps
Specifications
Specifications
| Quantity | Each |
| Type | Stand Clamp |
| Material | Zinc Die Cast, Powder Coated |
| Size (Metric) Grip | 12 to 25 mm |