missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZD 7288
GREENER_CHOICE

Product Code. 30001158
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001158 50 mg 50mg
18074612 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001158 Supplier Tocris Bioscience™ Supplier No. 1000/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits pacemaker (If) current

ZD 7288 is a sino-atrial node function modulator; blocks the hyperpolarization activated cation current If (HCN channel). Blocks Ih in central neurons. Increases NMDA-evoked noradrenaline release in rat brain in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133059-99-1
Molecular Formula C15H21ClN4

Specifications

Quantity 50 mg
Formula Weight 292.81
Purity >99%
Molecular Weight (g/mol) 292.811
Chemical Name or Material ZD 7288
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.