missing translation for 'onlineSavingsMsg'
Learn More

YO-01027, MedChemExpress

Product Code. p-200098933
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Product Code. Quantity unitSize
16485728 10 mg 10mg
16480196 2 mg 2mg
16428417 5 mg 5mg
16453366 25 mg 25mg
4 options
This item is not returnable. View return policy

Product Code. 16485728

Brand: Medchem Express HY13526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YO-01027 (Dibenzazepine;DBZ) is a potent γ-secretase inhibitor with IC50 values of 2.92 and 2.64 nM for Notch and APPL cleavage, respectively.

YO-01027 (Dibenzazepine

Specifications

Chemical Name or Material YO-01027
CAS 209984-56-5
Color White
Purity Grade Notes Research
Molecular Formula C26H23F2N3O3
Quantity 10 mg
Synonym Dibenzazepine DBZ
Solubility Information DMSO : 50 mg/mL (107.88 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N[C@@H](C)C(N[C@H]1C2=CC=CC=C2C3=CC=CC=C3N(C)C1=O)=O)CC4=CC(F)=CC(F)=C4
Molecular Weight (g/mol) 463.48
Formula Weight 463.48
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.