missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XEN723, MedChemExpress
Description
XEN723 is a novel and potent thiazolylimidazolidinone inhibitor of Stearoyl-CoA Desaturase (SCD1) with IC50s of 45 and 524 nM in mouse and HepG2 cell, respectively.
Specifications
Specifications
| Chemical Name or Material | XEN723 |
| CAS | 1072803-08-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20FN5O2S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (235.03 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(C)N=C(N2CCN(CC3=CC=C(F)C=C3)C2=O)S1)NCC4=CC=CN=C4 |
| Molecular Weight (g/mol) | 425.48 |
| Formula Weight | 425.48 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?