missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH130 C18 Column

Product Code. 13195179
Encompass
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
10 μm
Unit Size:
Each
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
13195179 50 mm 2.1 mm 3.5 μm Each
13125199 250 mm 2.1 mm 5 μm Each
13165179 50 mm 1 mm 3.5 μm Each
13175179 100 mm 1 mm 3.5 μm Each
13185179 150 mm 1 mm 3.5 μm Each
10044016 100 mm 2.1 mm 3.5 μm Each
13115189 150 mm 2.1 mm 3.5 μm Each
13125189 250 mm 2.1 mm 3.5 μm Each
13135189 50 mm 4.6 mm 3.5 μm Each
13145189 100 mm 4.6 mm 3.5 μm Each
10758117 150 mm 4.6 mm 3.5 μm Each
10565344 250 mm 4.6 mm 3.5 μm Each
13165189 50 mm 1 mm 5 μm Each
13175189 100 mm 1 mm 5 μm Each
13185189 150 mm 1 mm 5 μm Each
13195189 50 mm 2.1 mm 5 μm Each
13105199 100 mm 2.1 mm 5 μm Each
13115199 150 mm 2.1 mm 5 μm Each
13135199 50 mm 4.6 mm 5 μm Each
13145199 100 mm 4.6 mm 5 μm Each
13155199 150 mm 4.6 mm 5 μm Each
13165199 250 mm 4.6 mm 5 μm Each
13145259 50 mm 4.6 mm 10 μm Each
13155259 100 mm 4.6 mm 10 μm Each
13165259 150 mm 4.6 mm 10 μm Each
13175259 250 mm 4.6 mm 10 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
This item is not returnable. View return policy
Product Code. 13195179 Supplier Waters Corporation Supplier No. 186003563

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations.

  • The small-pore (130 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 130 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
Type Column
For Use With (Application) Peptides
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.