missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge BEH C18 XP Column

Product Code. 17172482
Encompass
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
75 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Quantity:
1/Ea.
3/Pk.
Unit Size:
Each
Pack of 3
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Quantity unitSize
17172482 75 mm 2.1 mm 3/Pk. Pack of 3
17112492 100 mm 3 mm 3/Pk. Pack of 3
15457174 100 mm 4.6 mm 1/Ea. Each
17162482 30 mm 2.1 mm 3/Pk. Pack of 3
14834521 50 mm 4.6 mm 1/Ea. Each
14804521 50 mm 3 mm 1/Ea. Each
15474881 50 mm 2.1 mm 1/Ea. Each
15651622 50 mm 2.1 mm 3/Pk. Pack of 3
17183432 100 mm 2.1 mm 1/Ea. Each
16631543 75 mm 4.6 mm 1/Ea. Each
17103442 75 mm 3 mm 1/Ea. Each
17182482 100 mm 2.1 mm 3/Pk. Pack of 3
17192482 30 mm 3 mm 3/Pk. Pack of 3
15890016 50 mm 3 mm 3/Pk. Each
17102492 75 mm 3 mm 3/Pk. Pack of 3
17163432 30 mm 2.1 mm 1/Ea. Each
17173432 75 mm 2.1 mm 1/Ea. Each
17193432 30 mm 3 mm 1/Ea. Each
17123442 30 mm 4.6 mm 1/Ea. Each
17142592 150 mm 2.1 mm 3/Pk. Pack of 3
17152592 150 mm 3 mm 3/Pk. Pack of 3
17113442 100 mm 3 mm 1/Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Product Code. 17172482 Supplier Waters Corporation Supplier No. 176002548

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12).

  • The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 75 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 3/Pk.
USP Type L1
Endcapped Yes
Type eXtended Performance (XP) Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.