Learn More
Kartell SpA Volumetric Flask
Product Code. 10051091
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
Unit Size:
Pack of 10
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10051091 50 mL Pack of 20
10172801 250 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10051091 Supplier Kartell SpA Supplier No. 0148500
Only null left
Add to basket
Add to basket
Volumetric flasks with cap, polypropylene
These flasks create no meniscus so final readings are easy. Calibrated gravimetrically at 20°C. Translucency of polypropylene enhanced when in contact with liquid. Thick walls eliminate distortion. Thin neck make final reading precise. Autoclavable. Chemically non-absorbant. No wetting. Stoppers included.
Specifications
Capacity (Metric) 50 mL
Material Polypropylene
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Threaded
Sterility Non-sterile
Bottom Shape Round
Surface Treatment Untreated