Learn More
Thermo Scientific™ Nalgene™ Staining Box
GREENER_CHOICE
Product Code. 10172491
Change view
Click to view available options
Type:
Gel Staining Box
Staining Box Plug
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10172491 Staining Box Plug Pack of 12
10492491 Gel Staining Box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10172491 Supplier Thermo Scientific™ Supplier No. 7157050001
Only null left
Add to basket
Add to basket
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods with these staining boxes.
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods. Thermo Scientific™ Nalgene™ Staining Box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 4.8 in. x 4.8 in (189 mm x 189 mm) gel along with 500mL of liquid.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a -70° to +70°C (-94° to +158°F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement Plug for 5705-1010 and -2020
Type Staining Box Plug
No. per Case 12
No. per Pack 12