Learn More
Waters Spherisorb SCX Column, 80Å, 5µm, 4.6mm X 250mm
Brand: Waters PSS837515
Spherisorb SCX Column, 80Å, 5µm, 4.6mm X 250mm
Safety and Handling
- Spherisorb SCX Column, 80Å, 5
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see supplemental first aid instructions on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use water spray to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contain: methanol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.