Learn More
Draeger Safety X-Plore 4790 Silicone Body
Product Code. 10143783
Change view
Click to view available options
:
Product Type: Half Mask
Product Code.
10143783 Product Type: Half Mask
1 options
Product Code. 10143783 Supplier Draeger Safety Supplier No. R55877
Currently Unavailable: We have no information when this product will be available.
View alternative products
Draeger Safety X-Plore 4790 Silicone Body is a half face mask, designed for use with the Rd90 filter range.
  • FlexiFit head harness means the mask can be donned and doffed quickly
  • Special harness helps to prevent hair entanglement and unnecessary pressure
  • Offers a tight fit to the face and leak tightness
  • Specially shaped nose seal and plastic frame add to a tight fit
Specifications
Certifications/Compliance CE certified (EN 140), AS/NZS
Connection Type Draeger-specific thread connection Rd90 (Draeger X-plore 4790)
Product Type Half Mask
Includes X-plore 4790 - size medium/large