missing translation for 'onlineSavingsMsg'
Learn More

WZ-3146, MedChemExpress

Product Code. p-200099518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447657 5 mg 5mg
16421157 50 mg 50mg
16414908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16447657 Supplier Medchem Express Supplier No. HY12001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WZ3146 is a mutant selective EGFR inhibitor with IC50s of 2, 2, 5, 14 and 66 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750, EGFRE746_A750/T790M and EGFR, respectively.

WZ3146 is a mutant selective EGFR inhibitor with IC50s of

Specifications

Chemical Name or Material WZ-3146
CAS 1214265-56-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25ClN6O2
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (134.42 mM; ultrasonic and warming and heat to 60°C)
SMILES CN1CCN(C2=CC=C(C=C2)NC3=NC=C(C(OC4=CC(NC(C=C)=O)=CC=C4)=N3)Cl)CC1
Molecular Weight (g/mol) 464.95
Formula Weight 464.95
Percent Purity 99.11%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.