missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red

Product Code. 10052432 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Neck Finish No. per Case unitSize
10052432 Plastic dropping bottle 8-425 1000 Cs. Pack of 1000
12446151 Dropping bottle 13-425 1000 Cs. Pack of 1000
12446131 Dropping bottle 20-410 1000 Cs. Pack of 1000
12436111 Dropping bottle 15-415 1000 Cs. Pack of 1000
10283673 Plastic dropping bottle 8-425 100 Cs. Pack of 100
10689293 Plastic dropping bottle 15-415 100 Cs. Pack of 100
11744189 Plastic dropping bottle 20-410 100 Cs. Pack of 100
10189102 Plastic dropping bottle 13-425 100 Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10052432 Supplier DWK Life Sciences Supplier No. W242502A
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 8-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.