missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
Shop All Wheaton Science Products ProductsDescription
Ideal for solvent, chemical and sample storage
- Narrow mouth enables liquids to be carefully poured to prevent spillage
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
- Available in partitioned Convenience or Valu-Bulk™ cartons
Order Info
Caps can be ordered separately.
Compliance
USP Type-III soda-lime glass
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Clear |
| Diameter (English) | 2.95 in. |
| Material | Soda Lime Glass |
| Packaging | Convenience pack |
| Closure Color | White |
| Certifications/Compliance | EPA, FDA |
| Height (Metric) | 168 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?