missing translation for 'onlineSavingsMsg'
Learn More

VSIG10 Antibody (988622) [DyLight 488], Novus Biologicals™

Product Code. 30761852 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761852 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761852 Supplier Novus Biologicals Supplier No. FAB9969K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSIG10 Monoclonal antibody specifically detects VSIG10 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen VSIG10
Applications Immunocytochemistry
Classification Monoclonal
Clone 988622
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias FLJ20674, V-set and immunoglobulin domain containing 10, V-set and immunoglobulin domain-containing protein 10
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human VSIG10, Met1-Gly413, Accession # Q8N0Z9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54621
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.