Learn More
Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™
Description
- Duran™ borosilicate glass 3.3.
- Fused blue graduation.
- Standard ground without stopper.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Capacity (Metric) | 200 mL |
| Material | Borosilicate Glass (3.3) |
| Neck Style | Standard |
| Product Type | Volumetric Flask |
| Graduated | Yes |
| Closure Size | NS 14 |