Learn More
Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™
Product Code. 10053801
Change view
Click to view available options
Capacity (Metric):
200 mL
250 mL
1000 mL
2000 mL
Closure Size:
NS 14
NS 24
NS 29
Neck Style:
Standard
Wide
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Neck Style unitSize
10053801 1000 mL NS 24 Standard Each
10165531 200 mL NS 14 Standard Each
10388971 250 mL NS 14 Standard Each
10516162 1000 mL NS 29 Wide Each
10319121 2000 mL NS 29 Standard Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10053801 Supplier Karl Hecht™ Supplier No. 42097080
Only null left
Add to basket
Add to basket
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask, Conformity Certified, Duran™ is thick-walled, clearly marked and labeled, and has a solid finish.
  • Duran™ borosilicate glass 3.3.
  • Fused blue graduation.
  • Standard ground without stopper.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications
Certifications/Compliance DIN EN ISO 1042
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
Neck Style Standard
Product Type Volumetric Flask
Graduated Yes
Closure Size NS 24