missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vivid™ CYP3A5 Green Screening Kit

Product Code. 10164792
Change view
Click to view available options
Quantity:
>300 assay(s)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10164792 >300 assay(s) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164792 Supplier Thermo Scientific™ Supplier No. P2969

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition.

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. They provide an optimized method for studying isozyme-specific CYP450-drug interactions, metabolism, and inhibition.

Vivid™ CYP450 Screening Kits offer:

  • Easy three-step procedure, 'mix and read' format, reactions at room temperature or 37°C
  • Superior fluorescent properties and kinetics compared to conventional fluorogenic probes
  • High signal-to-background ratio, broad dynamic range
  • Compatible with multiple assay formats from 96-well to 1536-well

Simple Mix and Read Kit Format

Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagents. The CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme (CYP3A5 in this case) and human cytochrome-P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

Unique Vivid™ Reagents for Bright Fluorescent Signals and Low Background

Vivid™ Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product (Figure 1). They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. Vivid™ DBOMF Substrate is the substrate included in this particular kit. This substrate yields a product that emits a green fluorescence.

Flexible Assay Formats for Optimized Results

The sensitivity of the Vivid™ CYP450 assays allows detection of weak inhibitors and miniaturization to as little as 2 μl per reaction. Assays may be set up in kinetic mode or in end-point mode to facilitate multi-plate screening (Figure 2). Assays may be performed at room temperature or 37°C.

Applications

  • High-throughput screening of enzyme-drug interactions
  • Compound profiling for drug inhibition of cytochrome P450 isozymes
  • Generation of predictive SAR models to guide compound acquisition
TRUSTED_SUSTAINABILITY

Specifications

Description Vivid™ CYP3A5 Green Screening Kit, 300 Assays, Liquid, Green Color, 96-well Plate Format, Fluorescent Detection Method, CYP3A5 Detection Target, VIVID™ Green Label, Use With Microplate Reader, Used in Cell Viability, Proliferation and Function, Pharma and Biopharma, Protein Assays and Analysis, Target-Based ADME/Tox Assays
For Use With (Equipment) Microplate Reader
Product Line VIVID
Formulation Liquid
For Use With (Application) Protein biology, cell analysis, cell viability, proliferation and function, cell metabolism, drug discovery and development, target-based adme and frasltox assays, biochemical ADME assays, cytochrome p450 inhibition adme assays, enzyme and protein activity assays, protein assays and analysis, cytochrome p450 activity
Includes Vivid™ CYP450 fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system
Content And Storage • Vivid™ CYP450 Fluorogenic substrates
• BACULOSOMES™ CYP450 reagents
• Fluorescent standards
• NADPH-regeneration system.

Vivid™ CYP450 Fluorogenic substrates are also available separately.

Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Detection Method Fluorescence
Label or Dye VIVID Green
Product Type Screening Kit
Quantity >300 assay(s)
Format 96-well Plate
Color Green
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.