missing translation for 'onlineSavingsMsg'
Learn More

3M™ Visor Protection Film

Product Code. 15819654
Change view
Click to view available options
:
Color: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15819654 Color: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819654 Supplier 3M™ Supplier No. 7100080225

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3M™ Visor Protection Film is a replacement part for use with 3M™ Protective Visors.

  • Peel-off protection film
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Equipment) Protective Visor
Product Type Visor Protection Film
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.