missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Oxoid™ Violet red bile agars powder

Product Code. 10036352
This item is not returnable. View return policy
Product Code. 10036352 Supplier Thermo Scientific™ Oxoid™ Supplier No. CM0485T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Appearance Straw/pink powder
Form Powder
For Use With (Application) Presumptive enumeration of Enterobacteriaceae
pH 7.2 to 7.6
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.