Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Straight
Description
- With straight side arms and hollow glass plug
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
Specifications
| Plug Bore | 8 mm |
| Material | Borosilicate Glass (3.3) |