Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Parallel
Description
- Acc. to DIN 12545 and in addition to DIN standard
- With parallel side arms and hollow glass plug
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
Specifications
| Plug Bore | 8 mm |
| Material | Borosilicate Glass (3.3) |