missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Universal Yellow 1-250 μL Pipette Tips

Product Code. 10156230
Click to view available options
Volume (Metric):
1 to 250 μL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 10156230

Brand: Ratiolab™ 2500160

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for all commercially available pipettes, both manual or electronic, single or multichannel.

  • Uniform high quality and precision makes these tips fully effective
  • Can be used with specially developed Multirack, Refill Systems, and Multiboxes
  • Provides for simple and safe pipetting
  • Can be used in the most sensitive applications
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Minimal residual wetting due to the controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Provided in packs of 1000

Specifications

Volume (Metric) 1 to 250 μL
Color Yellow
Autoclavable Autoclavable
Format Bulk
Tip Style Universal
Quantity 1000/pk
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.