missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Universal Pipette Filler

Product Code. 10145341
Change view
Click to view available options
:
For pipettes up to 100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10145341 For pipettes up to 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10145341 Supplier Karl Hecht™ Supplier No. 42581010
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For measuring and volumetric pipettes with capacities up to 100mL.

  • With 3 valves
  • With extended attachment
  • Made of red natural rubber

Specifications

Description With extended attachment
Color Red
Material Natural Rubber
Type Pipette Aid
For Use With (Equipment) Meauring and volumetric pipettes up to 100 mL capacity
Includes 3 valves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.