missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Universal Pipet Tips

Product Code. 10164964
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10164964 1 to 200 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10164964 Supplier Fisherbrand™ Supplier No. 2113602

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use these pipet tips with all commercially available pipets. Fisherbrand™ Universal Pipet Tips feature uniform high quality and precision for simpler, safer pipetting. Ideal for manual, electric and single- or multi-channel pipets.

  • High quality yellow polypropylene construction
  • Hydrophobic PE-HD filters provide an aerosol barrier and protect against contamination
  • Precisely fitting cone
  • Autoclavable
  • Stable shape allows for optimal handling
  • Excellent volume accuracy
  • Practice-oriented tip volumes

Specifications

Description Universal yellow pipet tip “E“
Volume (Metric) 1 to 200 μL
Color Yellow
Length (Metric) 4.95cm
Format Rack
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.