missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Union Nut

Product Code. 10644893
Change view
Click to view available options
Unit Size:
Each
Color:
Red
Turquoise
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Color
10644893 Each -
10004732 Each Turquoise
10710654 Each Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10644893 Supplier Hirschmann™ Supplier No. 9902402
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Retainer rings for use with the Pipetus™ pipette filler line.

  • Available in a range of colors
  • Aids in liquid handling
  • Accessory item

Specifications

Type Union Nut
For Use With (Equipment) Pipetus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.